Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFX1 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149932G
Description
NFX1 Polyclonal antibody specifically detects NFX1 in Human samples. It is validated for Western BlotSpecifications
NFX1 | |
Polyclonal | |
Western Blot | |
DKFZp779G2416, EC 6.3.2.-, MGC20369, NFX2transcriptional repressor NF-X1, Nuclear transcription factor, X box-binding protein 1, nuclear transcription factor, X-box binding 1 | |
The immunogen for this product maps to a region between residue 250 and 300 of human Nuclear Transcription Factor, X-box Binding 1 using the numbering given in entry NP_002495.2 (GeneID 4799). | |
0.1 mL | |
Zinc Finger | |
4799 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction