Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567299
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567299 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567299 Supplier Invitrogen™ Supplier No. PA567299
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSPPPCHLSRQ VPYDEISAVH QHSYHPSGSK PKSQQTSFQS SPCNKSPKSH GLQNQPWQKL RNEKHHIRVK KAQSLAEQTS DTA Highest antigen sequence identity to the following orthologs - mouse 74%, rat 74%.

MHC class II gene expression is controlled primarily at the transcriptional level by transcription factors that bind to the X and Y boxes, two highly conserved elements in the proximal promoter of MHC class II genes. NFX1 is a transcriptional repressor capable of binding to the conserved X box motif of HLA-DRA and other MHC class II genes in vitro. The protein may play a role in regulating the duration of an inflammatory response by limiting the period in which class II MHC molecules are induced by IFN-gamma.MHC class II gene expression is controlled primarily at the transcriptional level by transcription factors that bind to the X and Y boxes, two highly conserved elements in the proximal promoter of MHC class II genes. The protein encoded by this gene is a transcriptional repressor capable of binding to the conserved X box motif of HLA-DRA and other MHC class II genes in vitro. The protein may play a role in regulating the duration of an inflammatory response by limiting the period in which class II MHC molecules are induced by IFN-gamma. Three alternative splice variants, each of which encodes a different isoform, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFX1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NFX1
Gene Accession No. Q12986
Gene Alias 1300017N15Rik; 3000003M19Rik; DKFZp779G2416; MGC20369; m-Nfx.1; NFX.1; Nfx1; NFX2; nuclear transcription factor, X box-binding protein 1; nuclear transcription factor, X-box binding 1; TEG-42; testis expressed gene 42; Tex42; transcriptional repressor NF-X1; X-box binding 1
Gene Symbols NFX1
Host Species Rabbit
Immunogen Recombinant Human NFX1. Recombinant protein control fragment (Product #RP-107937).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4799
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.