Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFXL1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501564512
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4512 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4512 Supplier Bethyl Laboratories, Inc Supplier No. A303196AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

NFXL1 gene ontology annotations related to this gene include DNA-binding transcription factor activity; zinc ion binding.

Specifications

Antigen NFXL1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene NFXL1
Gene Accession No. Q6ZNB6
Gene Alias CDZFP, cytoplasm-distribution zinc finger protein, HOZFP, nuclear transcription factor, X-box binding-like 1, ovarian zinc finger protein, OZFP, up-regulated in colon cancer 5, URCC5
Gene Symbols NFXL1
Host Species Rabbit
Immunogen Between 100 and 150
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152518
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.