Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFYA Antibody, Novus Biologicals™
SDP

Catalog No. NB600481 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600481 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600481 Supplier Novus Biologicals Supplier No. NB600481
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NFYA Polyclonal specifically detects NFYA in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Gel Super Shift Assays.

Specifications

Antigen NFYA
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Gel Shift
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, ELISA 1:20000-1:100000, Immunohistochemistry 1:400, Immunohistochemistry-Paraffin 1:10-1:500, Gel Super Shift Assays 2.0 ul
Formulation Antiserum with 0.01% Sodium Azide
Gene Accession No. P23511
Gene Alias CAAT box DNA-binding protein subunit A, CBF-A, CBF-B, CCAAT-binding transcription factor subunit B, FLJ11236, HAP2, HAP2 CCAAT-binding protein, NF-YACAAT-box DNA binding protein subunit A, Nuclear transcription factor Y subunit A, nuclear transcription factor Y subunit alpha, nuclear transcription factor Y, alpha, Transcription factor NF-Y, A subunit
Gene Symbols NFYA
Host Species Rabbit
Immunogen NFYA (A subunit specific) peptide corresponding to a region near the N-terminus of the human protein conjugated to Keyhole Limpet Hemocyanin (KLH). (Uniprot: P23511)
Molecular Weight of Antigen 36.88 kDa
Purification Method Delipidation and Defibrination
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4800
Test Specificity Recognizes the A subunit of human and mouse NFY. No reaction was observed against the B subunit of NFY.
Target Species Human, Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.