Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFYA Antibody (SR1622), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32233950UL
Description
NFYA Monoclonal antibody specifically detects NFYA in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
NFYA | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
SR1622 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
CAAT box DNA-binding protein subunit A, CBF-A, CBF-B, CCAAT-binding transcription factor subunit B, FLJ11236, HAP2, HAP2 CCAAT-binding protein, NF-YACAAT-box DNA binding protein subunit A, Nuclear transcription factor Y subunit A, nuclear transcription factor Y subunit alpha, nuclear transcription factor Y, alpha, Transcription factor NF-Y, A subunit | |
A synthesized peptide derived from human NFYA (Uniprot #: P23511) | |
50 μg | |
Cancer, Signal Transduction, Transcription Factors and Regulators | |
4800 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction