Learn More
Invitrogen™ NFYA Polyclonal Antibody

Description
Recommended positive controls: A431, HeLa, HepG2, NIH-3T3, JC, BCL-1, PC-12, Rat2. Predicted reactivity: Mouse (99%), Rat (98%), Pig (97%), Rhesus Monkey (91%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | NFYA |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.19 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | NFYA |
| Gene Accession No. | P18576, P23511, P23708 |
| Gene Alias | AA407810; CAAT box DNA-binding protein subunit A; CAAT-box DNA binding protein subunit A; CAAT-box DNA-binding protein subunit A; CBF-A; Cbf-b; CCAAT-binding transcription factor subunit A; CCAAT-binding transcription factor subunit B; FLJ11236; HAP2; HAP2 CCAAT-binding protein; nf-y; NFYA; NF-YA; Nuclear transcription factor Y subunit A; nuclear transcription factor Y subunit alpha; nuclear transcription factor Y, alpha; nuclear transcription factor-Y alpha; SEZ10; SEZ-10; Transcription factor NF-Y, A subunit |
| Gene Symbols | NFYA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.