Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFYB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101096
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101096 Supplier Invitrogen™ Supplier No. PA5101096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NFYB.

The CCAAT-binding factor NF-Y is a heteromeric transcription factor that specifically binds to CCAAT sequences in many eukaryotic genes. NF-Y is made up of three subunits, NF-YA, NF-YB and NF-YC. All three components are necessary for DNA binding. In each NF-Y subunit, the segment needed for formation of the NF-Y-DNA complex is conserved from yeast to human. These conserved segments are homologous to the histone-fold motif of eukaryotic histones. The DNA-binding domains of the NF-YB and NF-YC subunits have been suggested to interact through a protein-protein, histone-fold handshake motif in a manner analogous to the histone proteins H2B and H2A, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFYB
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NFYB
Gene Accession No. P25208, P63139, P63140
Gene Alias AA985999; CAAT box DNA-binding protein subunit B; CAAT-box DNA-binding protein subunit B; CBF-A; CBFB; CBF-B; CCAAT binding transcription factor of CBF-B/NFY-B; CCAAT-binding transcription factor subunit A; CCAAT-binding transcription factor subunit B; HAP3; nf-y; Nfya; Nfyb; NF-YB; nuclear transcription factor - Y beta; nuclear transcription factor Y subunit B; Nuclear transcription factor Y subunit beta; nuclear transcription factor Y, beta; nuclear transcription factor-Y beta; Transcription factor NF-Y, B subunit
Gene Symbols NFYB
Host Species Rabbit
Immunogen A synthesized peptide derived from human NFYB(Accession P25208), corresponding to amino acid residues Q14-V64.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18045, 25336, 4801
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.