Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFYC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541548
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541548 Supplier Thermo Scientific Supplier No. PA541548
Add to Cart
Add to Cart

NFYC Polyclonal Antibody

NFYC Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen NFYC
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NFYC
Gene Accession No. P70353, Q13952
Gene Alias CAAT box DNA-binding protein subunit C; CAAT-box DNA-binding protein subunit C; CBFC; CBF-C; CCAAT binding factor of CBF-C/NFY-C; CCAAT binding factor subunit C; CCAAT transcription binding factor subunit gamma; CCAAT-binding transcription factor subunit C; DKFZp667G242; fb98f01; FLJ45775; H1TF2A; HAP5; histone H1 transcription factor large subunit 2A; HSM; NFYC; NF-YC; Nuclear transcription factor Y subunit C; nuclear transcription factor Y subunit gamma; nuclear transcription factor Y, gamma; nuclear transcription factor-Y gamma; transactivator HSM-1; transactivator HSM-1/2; transcription factor NF-Y, C subunit; wu:fb98f01; zgc:55514
Gene Symbols NFYC
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human NFYC.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18046, 4802
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.