Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFYC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565917
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565917 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565917 Supplier Invitrogen™ Supplier No. PA565917
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPVQYYFTLAQ QPTAVQVQGQ QQGQQTTSST TTIQPGQIII AQPQQGQTTP VTMQVGEGQQ VQIVQAQPQG QAQQAQSGTG QTMQVMQQII TNTGEIQQIP VQLNAGQLQY IRLAQPVSGT QVVQGQIQTL ATNAQ Highest antigen sequence identity to the following orthologs - mouse 99%, rat 100%.

The CCAAT-binding factor NF-Y is a heteromeric transcription factor that specifically binds to CCAAT sequences in many eukaryotic genes. NF-Y is made up of three subunits, NF-YA, NF-YB and NF-YC. All three components are necessary for DNA binding. In each NF-Y subunit, the segment needed for formation of the NF-Y-DNA complex is conserved from yeast to human. These conserved segments are homologous to the histone-fold motif of eukaryotic histones. The DNA binding domains of the NF-YB and NF-YC subunits have been suggested to interact through a protein-protein histone-fold 'handshake' motif in a manner analogous to the histone proteins, H2B and H2A, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFYC
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NFYC
Gene Accession No. Q13952
Gene Alias CAAT box DNA-binding protein subunit C; CAAT-box DNA-binding protein subunit C; CBFC; CBF-C; CCAAT binding factor of CBF-C/NFY-C; CCAAT binding factor subunit C; CCAAT transcription binding factor subunit gamma; CCAAT-binding transcription factor subunit C; DKFZp667G242; fb98f01; FLJ45775; H1TF2A; HAP5; histone H1 transcription factor large subunit 2A; HSM; NFYC; NF-YC; Nuclear transcription factor Y subunit C; nuclear transcription factor Y subunit gamma; nuclear transcription factor Y, gamma; nuclear transcription factor-Y gamma; transactivator HSM-1; transactivator HSM-1/2; transcription factor NF-Y, C subunit; wu:fb98f01; zgc:55514
Gene Symbols NFYC
Host Species Rabbit
Immunogen Recombinant Human NFYC. Recombinant protein control fragment (Product #RP-106594).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4802
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.