Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFYC Polyclonal Antibody

NFYC Polyclonal Antibody
Supplier: Thermo Scientific PA5106696
Description
NFYC Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| NFYC | |
| Polyclonal | |
| Unconjugated | |
| NFYC | |
| CAAT box DNA-binding protein subunit C; CAAT-box DNA-binding protein subunit C; CBFC; CBF-C; CCAAT binding factor of CBF-C/NFY-C; CCAAT binding factor subunit C; CCAAT transcription binding factor subunit gamma; CCAAT-binding transcription factor subunit C; DKFZp667G242; fb98f01; FLJ45775; H1TF2A; HAP5; histone H1 transcription factor large subunit 2A; HSM; NFYC; NF-YC; Nuclear transcription factor Y subunit C; nuclear transcription factor Y subunit gamma; nuclear transcription factor Y, gamma; nuclear transcription factor-Y gamma; transactivator HSM-1; transactivator HSM-1/2; transcription factor NF-Y, C subunit; wu:fb98f01; zgc:55514 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18046, 25337, 4802 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P70353, Q13952, Q62725 | |
| NFYC | |
| A synthesized peptide derived from human NFYC(Accession Q13952), corresponding to amino acid residues A65-I115. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction