Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NG2 Chondroitin Sulfate Proteoglycan Mouse anti-Rat, Mouse, Unconjugated, Clone: 132.38, MilliporeSigma™

Catalog No. MAB5384I100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5384-I-100 100 μg
MAB5384-I-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB5384-I-100 Supplier MilliporeSigma Supplier No. MAB5384I100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-NG2 Chondroitin Sulfate Proteoglycan, clone 132.38, Cat. No. MAB5384-I, is a highly specific mouse monoclonal antibody that targets Chondrin Sulfate Protoglycan 4 and has been tested for use in Immunofluorescence and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NG2 Chondroitin Sulfate Proteoglycan
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 132.38
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing PBS, pH 7.2, containing 0.05% sodium azide.
Gene Alias Chondroitin sulfate proteoglycan 4;Chondroitin sulfate proteoglycan NG2;HSN tumor-specific antigen;NG2
Gene Symbols Cspg4;Ng2
Host Species Mouse
Immunogen HEK293 cells expressing a truncated integral membrane form of NG2 consisting of amino acids 1592-2222.
Molecular Weight of Antigen ∽260 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_112284
Test Specificity Clone 132.38 is a mouse monoclonal antibody that detects Chondroitin sulfate proteoglycan 4 in rat brain tissue.
Target Species Rat, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.