Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NG2/MCSP Antibody (LHM 2), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002688PECY55
Description
NG2/MCSP Monoclonal specifically detects NG2/MCSP in Human samples. It is validated for Flow Cytometry, Knockout Validated.Specifications
NG2/MCSP | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
CSPG4 | |
A 375P cells crude extract | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry, Immunoassay | |
LHM 2 | |
Flow Cytometry, Knockout Validated | |
chondroitin sulfate proteoglycan 4, chondroitin sulfate proteoglycan 4 (melanoma-associated), EC 3.6.3, HMW-MAA, MCSPChondroitin sulfate proteoglycan NG2, MCSPGMelanoma chondroitin sulfate proteoglycan, MEL-CSPG, MSK16Melanoma-associated chondroitin sulfate proteoglycan, NG2EC 2.7.8 | |
Mouse | |
Protein G purified | |
Cancer, Neuroscience | |
1464.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction