Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NG2 Rat anti-Mouse, Unconjugated, Clone: 1E6, MilliporeSigma™

Catalog No. MABS19491MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS19491MI 100 μg
MABS19492MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS19491MI Supplier MilliporeSigma Supplier No. MABS1949100UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

NG2 Monoclonal antibody specifically detects NG2 in Mouse samples. It is validated for ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NG2
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1E6
Conjugate Unconjugated
Gene Accession No. NP_620570
Gene Alias Chondroitin sulfate proteoglycan 4;Chondroitin sulfate proteoglycan NG2;Proteoglycan AN2
Gene Symbols Cspg4, An2, Kiaa4232, Ng2
Host Species Rat
Immunogen Oli-Neu cells (murine oligodendroglial precursor cells immortalized by an activated neu tyrosine kinase).
Molecular Weight of Antigen ∼330 kDa observed; 252.31 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 1E6 is a rat monoclonal antibody that detects mouse Chondroitin sulfate proteoglycan 4 (NG2).
Target Species Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.