Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGAL (LCN2) Monoclonal Antibody (OTI2E1), TrueMAB™, OriGene
SDP

Catalog No. 502543329
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3329 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3329 Supplier OriGene Supplier No. CF814415
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NGAL (LCN2) Monoclonal antibody specifically detects NGAL (LCN2) in Human samples. It is validated for ELISA

Specifications

Antigen NGAL (LCN2)
Applications ELISA
Classification Monoclonal
Clone OTI2E1
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene LCN2
Gene Accession No. P80188
Gene Alias 24p3; 25 kDa alpha-2-microglobulin-related subunit of MMP-9; alpha-2-microglobulin-related protein; Alpha-2U globulin-related protein; AW212229; HGNC:6526; HNL; LCN2; lipocalin 2; lipocalin 2 (oncogene 24p3); Lipocalin 24p3; Lipocalin2; Lipocalin-2; migration-stimulating factor inhibitor; MSFI; neutrophil gelatinase-associated lipocalin; NGAL; oncogene 24p3; OTTHUMP00000022240; p25; RP23-161B9.11; secreted inducible protein 24; Siderocalin; Siderocalin LCN2; Sip24; SV-40-induced 24P3 protein; uterocalin
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 21-198 of human NGAL (NP_005555) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3934
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.