Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NGAL (LCN2) Monoclonal Antibody (OTI4G10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF814368


Description
NGAL (LCN2) Monoclonal antibody specifically detects NGAL (LCN2) in Human samples. It is validated for ELISASpecifications
NGAL (LCN2) | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P80188 | |
Mouse | |
Protein A/G | |
RUO | |
3934 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
ELISA | |
OTI4G10 | |
Unconjugated | |
LCN2 | |
24p3; 25 kDa alpha-2-microglobulin-related subunit of MMP-9; alpha-2-microglobulin-related protein; Alpha-2U globulin-related protein; AW212229; HGNC:6526; HNL; LCN2; lipocalin 2; lipocalin 2 (oncogene 24p3); Lipocalin 24p3; Lipocalin2; Lipocalin-2; migration-stimulating factor inhibitor; MSFI; neutrophil gelatinase-associated lipocalin; NGAL; oncogene 24p3; OTTHUMP00000022240; p25; RP23-161B9.11; secreted inducible protein 24; Siderocalin; Siderocalin LCN2; Sip24; SV-40-induced 24P3 protein; uterocalin | |
Full length human recombinant protein of human NGAL (NP_005555) produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction