Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NGAL Monoclonal Antibody (29)
GREENER_CHOICE

Catalog No. ABS0432902
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS0432902 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS0432902 Supplier Invitrogen™ Supplier No. ABS0432902
Add to Cart
Add to Cart

Rat Monoclonal Antibody

ABS 043-29-02 binds free NGAL in mouse serum. This antibody will not cross react with human, pig, dog, or rat. ABS 043-29-02 may be used in Western blot and ELISA applications. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

NGAL (Neutrophil gelatinase-associated lipocalin, lipocalin 2, siderocalin and neutrophil lipocalin) is a 25 kDa glycosylated single chain monomer and member of the lipocalin family of proteins that bind and transport small lipophilic molecules. NGAL is released by activated neutrophils, can form dimers and small amounts of higher oligomers, and complexes with matrix metalloproteinase 9 (MMP-9, gelatinase B). Low level expression of NGAL in a variety of epithelia may be increased in inflammation or cancers. Expression of NGAL in the kidney is dramatically increased by ischemic or nephrotoxic kidney injury.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NGAL
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 29
Concentration 0.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene LCN2
Gene Accession No. P11672
Gene Alias 24p3; 25 kDa alpha-2-microglobulin-related subunit of MMP-9; alpha-2-microglobulin-related protein; Alpha-2U globulin-related protein; AW212229; HGNC:6526; HNL; LCN2; lipocalin 2; lipocalin 2 (oncogene 24p3); Lipocalin 24p3; Lipocalin2; Lipocalin-2; migration-stimulating factor inhibitor; MSFI; neutrophil gelatinase-associated lipocalin; NGAL; oncogene 24p3; OTTHUMP00000022240; p25; RP23-161B9.11; secreted inducible protein 24; Siderocalin; Siderocalin LCN2; Sip24; SV-40-induced 24P3 protein; uterocalin
Gene Symbols LCN2
Host Species Rat
Immunogen Recombinant mouse NGAL adsorbed onto aluminum hydroxide gel.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16819
Target Species Mouse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.