Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NGBR/Nogo B receptor Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11468R

Description
With DHDDS, forms the dehydrodolichyl diphosphate synthase (DDS) complex, an essential component of the dolichol monophosphate (Dol-P) biosynthetic machinery. Adds multiple copies of isopentenyl pyrophosphate (IPP) to farnesyl pyrophosphate (FPP) to produce dehydrodolichyl diphosphate (Dedol-PP), a precursor of dolichol which is utilized as a sugar carrier in protein glycosylation in the endoplasmic reticulum (ER). Regulates the glycosylation and stability of nascent NPC2, thereby promoting trafficking of LDL-derived cholesterol. Acts as a specific receptor for the N-terminus of Nogo-B, a neural and cardiovascular regulator.Specifications
NGBR/Nogo B receptor | |
Polyclonal | |
Unconjugated | |
NUS1 | |
1600027K07Rik; AU019165; AW538011; BC003223; C6orf68; Cis-prenyltransferase subunit NgBR; D10Ertd438e; dehydrodolichyl diphosphate syntase complex subunit NUS1; Dehydrodolichyl diphosphate synthase complex subunit Nus1; di-trans,poly-cis-decaprenylcistransferase; MGC:7199; NGBR; Nogo-B receptor; Nuclear undecaprenyl pyrophosphate synthase 1 homolog; nuclear undecaprenyl pyrophosphate synthase 1 homolog (S. cerevisiae); NUS1; NUS1 dehydrodolichyl diphosphate synthase subunit; NUS1, dehydrodolichyl diphosphate synthase subunit; RGD1307879; TANGO14; transport and golgi organization 14 homolog | |
Rabbit | |
Protein A | |
RUO | |
116150 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q96E22 | |
NUS1 | |
KLH conjugated synthetic peptide derived from human NGBR/Nogo B receptor . | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction