Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NGDN Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318494
This item is not returnable.
View return policy
Description
NGDN Polyclonal antibody specifically detects NGDN in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
NGDN | |
Polyclonal | |
Unconjugated | |
Tris-citrate/phosphate pH 7 -8. | |
Rabbit | |
Affinity purified | |
RUO | |
25983 | |
Store at 4°C. Do not freeze. | |
IgG |
Western Blot, Immunoprecipitation | |
1 mg/ml | |
Western Blot 1:2000 - 1:10000, Immunoprecipitation 20-40 ug/mg lysate | |
C14orf120, DKFZp564O092, EIF4E-binding protein, eukaryotic initiation factor 4E and CPEB binding protein, LCP5, lpd-2, neuroguidin, neuroguidin, EIF4E binding protein, NGDchromosome 14 open reading frame 120 | |
Recognizes region between residues Between between 100 and 150.. | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction