Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NGDN Antibody (OTI1H4), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272979AF405
Description
NGDN Monoclonal specifically detects NGDN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
NGDN | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NGDN | |
Full length recombinant protein of human NGDN (NP_001036100) produced in E.coli. | |
0.1 mL | |
25983.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1H4 | |
Western Blot, Immunohistochemistry | |
C14orf120, DKFZp564O092, EIF4E-binding protein, eukaryotic initiation factor 4E and CPEB binding protein, LCP5, lpd-2, neuroguidin, neuroguidin, EIF4E binding protein, NGDchromosome 14 open reading frame 120 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction