Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NGDN Monoclonal Antibody (OTI1B8)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527059

Catalog No. PIMA527059


Only null left
Add to Cart

Description

Description

NGDN Monoclonal Antibody for Western Blot, IHC (P)

Involved in the translational repression of cytoplasmic polyadenylation element (CPE)-containing mRNAs.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NGDN
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8NEJ9
Ngdn
Full length human recombinant protein of NGDN produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1B8
Unconjugated
Ngdn
1500001L15Rik; C14orf120; CANu1; Centromere accumulated nuclear protein 1; EIF4E-binding protein; eukaryotic initiation factor 4E and CPEB binding protein; LCP5; lpd-2; neuroguidin; neuroguidin, EIF4E binding protein; NGD; Ngdn; RGD1310211
Mouse
Affinity Chromatography
RUO
25983
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.