Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NGDN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120276
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120276 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120276 Supplier Invitrogen™ Supplier No. PA5120276
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The target is usually found in the following locations: Cell projection, Chromosome, Cytoplasm, Nucleus, axon, centromere, dendrite, filopodium, nucleolus. Immunogen sequence: LMYLMDLTHL ILDKASGGSL QGHDAVLRLV EIRTVLEKLR PLDQKLKYQI DKLIKTAVTG SLSENDPLRF KPHPSNMMSK LSSEDEEEDE AEDDQSEASG KKSVKGVSKK YVPPRLVPVH YDETEAEREK KRLERAKRRA LSSSVIREL.

Involved in the translational repression of cytoplasmic polyadenylation element (CPE)-containing mRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NGDN
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ngdn
Gene Accession No. Q8NEJ9
Gene Alias 1500001L15Rik; C14orf120; CANu1; Centromere accumulated nuclear protein 1; EIF4E-binding protein; eukaryotic initiation factor 4E and CPEB binding protein; LCP5; lpd-2; neuroguidin; neuroguidin, EIF4E binding protein; NGD; Ngdn; RGD1310211
Gene Symbols Ngdn
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 61-209 of human NGDN (NP_001036100.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25983, 305887
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.