Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NGEF Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552521
Description
Immunogen sequence: METRESEDLE KTRRKSASDQ WNTDNEPAKV KPELLPEKEE TSQADQDIQD KEPHCHIPIK RNSIFNRSIR RKSKAKARDN PERNASCLAD SQDNGKSVNE PLTLNIPWSR TPPCRTAMQT DPGAQEMSES SST Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 26%.
Acts as a guanine nucleotide exchange factor (GEF) which differentially activates the GTPases RHOA, RAC1 and CDC42. Plays a role in axon guidance regulating ephrin-induced growth cone collapse and dendritic spine morphogenesis. Upon activation by ephrin through EPHA4, the GEF activity switches toward RHOA resulting in its activation. Activated RHOA promotes cone retraction at the expense of RAC1- and CDC42-stimulated growth cone extension.
Specifications
| NGEF | |
| Polyclonal | |
| Unconjugated | |
| NGEF | |
| ARHGEF27; Besh3; brain-enriched SH3-domain protein Besh3; ephexin; ephexin-1; eph-interacting exchange protein; Neuronal guanine nucleotide exchange factor; Ngef; Tims2; transforming immortalised mammary epithelial sequence 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25791 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N5V2 | |
| NGEF | |
| Recombinant protein corresponding to Human NGEF. Recombinant protein control fragment (Product #RP-89831). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction