Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGFI-B alpha/Nur77/NR4A1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056745T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056745T 0.025 mg
NB10056745 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056745T Supplier Novus Biologicals Supplier No. NB10056745SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 21 publication

NGFI-B alpha/Nur77/NR4A1 Polyclonal specifically detects NGFI-B alpha/Nur77/NR4A1 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen NGFI-B alpha/Nur77/NR4A1
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Chromatin Immunoprecipitation 1:20-1:1000. Use reported in scientific literature (Lee et al (2009)), Immunohistochemistry 1:200. Use reported in scientific literature (PMID 31115481), Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (Chintharlapalli et al (2005)), Immunoprecipitation 1:20-1:1000. Use reported in scientific literature (Ferlini et al (2009)), Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P22736
Gene Alias Early response protein NAK1, GFRP1ST-59, growth factor-inducible nuclear protein N10, HMRNP10, hormone receptor, MGC9485, N10, NAK1, NAK-1, NGFIB, Nuclear hormone receptor NUR/77, nuclear receptor subfamily 4 group A member 1, nuclear receptor subfamily 4, group A, member 1, NUR77, Orphan nuclear receptor HMR, Orphan nuclear receptor TR3, steroid receptor TR3, Testicular receptor 3, TR3, TR3 orphan receptor
Gene Symbols NR4A1
Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to aa 251-266 of human Nak1 (NP_775180).
Purification Method Immunogen affinity purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 3164
Test Specificity Human and Mouse. Not yet tested in other species.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.