Learn More
Description
Specifications
Specifications
| Antigen | NGFI-B alpha/Nur77/NR4A1 |
| Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:1000, Chromatin Immunoprecipitation 1:20-1:1000. Use reported in scientific literature (Lee et al (2009)), Immunohistochemistry 1:200. Use reported in scientific literature (PMID 31115481), Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (Chintharlapalli et al (2005)), Immunoprecipitation 1:20-1:1000. Use reported in scientific literature (Ferlini et al (2009)), Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000 |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Accession No. | P22736 |
| Gene Alias | Early response protein NAK1, GFRP1ST-59, growth factor-inducible nuclear protein N10, HMRNP10, hormone receptor, MGC9485, N10, NAK1, NAK-1, NGFIB, Nuclear hormone receptor NUR/77, nuclear receptor subfamily 4 group A member 1, nuclear receptor subfamily 4, group A, member 1, NUR77, Orphan nuclear receptor HMR, Orphan nuclear receptor TR3, steroid receptor TR3, Testicular receptor 3, TR3, TR3 orphan receptor |
| Gene Symbols | NR4A1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
