Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NGFR Monoclonal Antibody (ME20.4), FITC
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA118420
Description
The antibody conjugate can be stored at 4 °C for up to 4 months with the addition of appropriate antibacterial agent.
Neurotrophic factors control the survival, differentiation and maintenance of neurons in the peripheral and central nervous systems, and of other neural crest-derived cell types. Developing sympathetic neurons are absolutely dependent upon nerve growth factor (NGF) during the period of target competition in vivo. Antibodies reacting specifically with NGFR p75 are useful tools in the detection and characterization of NGFR p75, to enhance our understanding of a wide range of phenomena in the development, plasticity and repair of the nervous system.
Specifications
NGFR | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P08138 | |
Ngfr | |
Human WM245 melanoma cells. | |
100 μg | |
Primary | |
Human, Canine, Rabbit, Sheep, Pig, Feline | |
Antibody | |
IgG1 |
Immunohistochemistry (Frozen), Immunocytochemistry | |
ME20.4 | |
FITC | |
Ngfr | |
CD271; CD271 antigen; Gp80-LNGFR; LNGFR; low affinity nerve growth factor receptor; low affinity neurotrophin receptor p75NTR; LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 16; low-affinity nerve growth factor receptor; nerve growth factor receptor; nerve growth factor receptor (TNFR superfamily, member 16); nerve growth factor receptor, fast; NGF receptor; NGFR; NGR; p75; p-75; p75 ICD; p75 neurotrophin receptor; p75 ntr; p75(NTR); p75NGFR; p75NTR; RNNGFRR; TNFR superfamily, member 16; Tnfrsf16; TNR16; Tumor necrosis factor receptor superfamily member 16 | |
Mouse | |
Protein G | |
RUO | |
100349095, 100525607, 101101519, 101119064, 4804, 491071 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction