Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NGFR/TNFRSF16/p75NTR Antibody (NGFR5 + NTR/912), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247967MFV610
Description
NGFR/TNFRSF16/p75NTR Monoclonal antibody specifically detects NGFR/TNFRSF16/p75NTR in Human, Primate, Mouse (Negative), Rat (Negative) samples. It is validated for Flow CytometrySpecifications
NGFR/TNFRSF16/p75NTR | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Primate, Mouse (Negative), Rat (Negative) | |
Purified |
Flow Cytometry | |
NGFR5 + NTR/912 | |
Flow Cytometry | |
CD271, CD271 antigen, Gp80-LNGFR, member 16, nerve growth factor receptor, NGF receptor, p75 ICD, TNFRSF16nerve growth factor receptor (TNFR superfamily, member 16), tumor necrosis factor receptor superfamily member 16 | |
NGFR from A875 melanoma cells (NGFR5); Recombinant human NGFR/TNFRSF16/p75NTR protein (NTR/912) | |
0.1 mL | |
Apoptosis, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
4804 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction