Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NGFR/TNFRSF16/p75NTR Antibody (SPM299), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | NGF R/TNFRSF16/p75NTR |
---|---|
Clone | SPM299 |
Concentration | 0.2mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24487500
![]() |
Novus Biologicals
NBP2448750.02MG |
0.02 mg |
Each for $268.00
|
|
|||||
NBP24487501
![]() |
Novus Biologicals
NBP2448750.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
NBP24487502
![]() |
Novus Biologicals
NBP2448750.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
NGFR/TNFRSF16/p75NTR Monoclonal specifically detects NGFR/TNFRSF16/p75NTR in Human, Feline, Ferret, Monkey, Baboon, Rabbit, Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
NGF R/TNFRSF16/p75NTR | |
0.2mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Mouse | |
Apoptosis, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
4804 | |
NGFR from A875 melanoma cells | |
Primary | |
Store at 4C. | |
75 kDa |
SPM299 | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Baboon, Feline, Ferret, Monkey, Rabbit, Mouse (Negative), Rat (Negative) | |
CD271, CD271 antigen, Gp80-LNGFR, member 16, nerve growth factor receptor, NGF receptor, p75 ICD, TNFRSF16nerve growth factor receptor (TNFR superfamily, member 16), tumor necrosis factor receptor superfamily member 16 | |
NGFR | |
IgG1 κ | |
Protein A or G purified | |
It recognizes a glycoprotein of 75kDa, identified as low affinity Nerve Growth Factor (NGF) Receptor (p75NGFR) or Neurotrophin Receptor (p75NTR). Its epitope spans in aa 1-160 of extracellular domain of NGFR/NTR. NGF-receptor contains an extracellular domain containing four 40-amino acid repeats with 6 cysteine residues at conserved positions followed by a serine/threonine-rich region, a single transmembrane domain, and a 155-amino acid cytoplasmic domain. The cysteine-rich region contains the nerve growth factor binding domain. NGF is important for the development, differentiation, and survival of variety of neuronal and non-neuronal cells. Its action is mediated by binding two distinct receptors, the high affinity p140 and low affinity p75. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title