Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGFRAP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987914
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7914 Supplier Bioss Supplier No. BS7671R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be a signaling adapter molecule involved in p75NTR-mediated apoptosis induced by NGF. Plays a role in zinc-triggered neuronal death (By similarity). May play an important role in the pathogenesis of neurogenetic diseases.

Specifications

Antigen NGFRAP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BEX3
Gene Accession No. Q00994
Gene Alias AL033356; Bex; BEX3; brain expressed X-linked 3; Brain-expressed X-linked protein 3; Brain-expressed X-linked protein 3 homolog; DXS6984E; DXWsu67e; Gcap27; granule cell antiserum positive 27; HGR74; Nade; nerve growth factor receptor (TNFRSF16) associated protein 1; nerve growth factor receptor associated protein 1; Nerve growth factor receptor-associated protein 1; NGFRAP1; Ovarian granulosa cell 13.0 kDa protein HGR74; p75NTR-associated cell death executor; Protein BEX3; rBex3; RP13-349O20.1
Gene Symbols BEX3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NADE/NGFRAP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27018
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.