Learn More
NGLY1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305547AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes an enzyme that catalyzes hydrolysis of an N(4)-(acetyl-beta-D-glucosaminyl) asparagine residue to N-acetyl-beta-D-glucosaminylamine and a peptide containing an aspartate residue. The encoded enzyme may play a role in the proteasome-mediated degradation of misfolded glycoproteins. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
NGLY1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q96IV0 | |
NGLY1 | |
Between 250 and 300 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
NGLY1 | |
Peptide-N(4)-(N-acetyl-beta-glucosaminyl)asparagine amidase; PNGase; hPNGase; Peptide:N-glycanase; N-glycanase 1; PNG1; NGLY1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55768 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.