Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ngp Polyclonal Antibody, Rockland™
SDP

Catalog No. 501992127
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2127 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-2127 Supplier Rockland Immunochemicals Supplier No. 600401GW9
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Ngp is directed against full-length mouse Ngp. A BLAST analysis was used to suggest reactivity with this protein in Ngp based on 100% homology match for the immunogen sequence.

A proteomic study in 2011 determined a direct correlation between NGP mRNA levels and myeloid-derived suppressor cells. Specifically cathepsin B, a catalytic mechanism and potentially biomarker in a variety of cancers and amyloid plaque development, was inhibited by NGP. The significance of NGP as a negative regulator of tumor metastasis makes this antibody ideal for investigators involved in cancer research, as well as Alzheimer's research.

Specifications

Antigen Ngp
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Ngp
Gene Accession No. O08692
Gene Symbols Ngp
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18054
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.