Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NgR2/NgRH1/RTN4RL2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB435957 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB435957 0.025 mg
NBP241200 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB435957 Supplier Novus Biologicals Supplier No. NBP2412000.025MG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NgR2/NgRH1/RTN4RL2 Polyclonal specifically detects NgR2/NgRH1/RTN4RL2 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen NgR2/NgRH1/RTN4RL2
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA
Formulation PBS with 0.02% Sodium Azide
Gene Alias NgR2, NGRH1, reticulon 4 receptor-like 2
Gene Symbols RTN4RL2
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human RTN4RL2. The immunogen is located within amino acids 330 - 380 of RTN4RL2. Amino Acid Squence: LYRDLPAEDSRGRQGGDA
Molecular Weight of Antigen 46 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 349667
Test Specificity Three alternatively spliced transcript variants have been observed. RTN4RL2 antibody is predicted to not cross-react with other Nogo receptor-like proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.