Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NgR2/NgRH1/RTN4RL2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241200
Description
NgR2/NgRH1/RTN4RL2 Polyclonal specifically detects NgR2/NgRH1/RTN4RL2 in Human samples. It is validated for Western Blot, ELISA.Specifications
NgR2/NgRH1/RTN4RL2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
RTN4RL2 | |
Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human RTN4RL2. The immunogen is located within amino acids 330 - 380 of RTN4RL2. Amino Acid Squence: LYRDLPAEDSRGRQGGDA | |
Affinity Purified | |
RUO | |
349667 | |
Human | |
IgG |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 1-2 μg/mL, ELISA | |
NgR2, NGRH1, reticulon 4 receptor-like 2 | |
Rabbit | |
46 kDa | |
0.1 mg | |
Primary | |
Three alternatively spliced transcript variants have been observed. RTN4RL2 antibody is predicted to not cross-react with other Nogo receptor-like proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction