Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NgR2/NgRH1/RTN4RL2 Antibody (P14) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | NgR2/NgRH1/RTN4RL2 |
---|---|
Clone | P14 |
Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25267625
![]() |
Novus Biologicals
NBP2526760.025MG |
0.025 mg |
Each for $268.00
|
|
|||||
NBP25267602
![]() |
Novus Biologicals
NBP2526760.2MG |
0.2 mg |
Each for $584.50
|
|
|||||
Description
NgR2/NgRH1/RTN4RL2 Monoclonal specifically detects NgR2/NgRH1/RTN4RL2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF-ready.Specifications
NgR2/NgRH1/RTN4RL2 | |
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
349667 | |
(His)6-tagged fragment of rat NgR2 (amino acids 279-420). | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
P14 | |
Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
NgR2, NGRH1, reticulon 4 receptor-like 2 | |
RTN4RL2 | |
IgG | |
Protein A or G purified | |
The antibody binds to human NgR2 with a Kd of 2.3 nM and to rat NgR2 with a Kd of 0.48 nM (bivalent binding). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title