Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NgR2/NgRH1/RTN4RL2 Antibody (P14), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252676MFV450
Description
NgR2/NgRH1/RTN4RL2 Monoclonal antibody specifically detects NgR2/NgRH1/RTN4RL2 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
NgR2/NgRH1/RTN4RL2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
P14 | |
Flow Cytometry | |
NgR2, NGRH1, reticulon 4 receptor-like 2 | |
(His)6-tagged fragment of rat NgR2 (amino acids 279-420). | |
0.1 mL | |
Cellular Signaling | |
349667 | |
Store at 4C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction