Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NgR3/NgRH2/RTN4RL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP309588100UL

 View more versions of this product

Catalog No. NB124077


Only null left
Add to Cart

Description

Description

NgR3/NgRH2/RTN4RL1 Polyclonal specifically detects NgR3/NgRH2/RTN4RL1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

NgR3/NgRH2/RTN4RL1
Polyclonal
Western Blot 1.0 ug/ml
DKFZp547J144, NgR3, NGRH2, NGRL2, nogo receptor-like 2, Nogo-66 receptor homolog 2, Nogo-66 receptor-related protein 3, reticulon 4 receptor-like 1, reticulon-4 receptor-like 1
The immunogen is a synthetic peptide directed towards the C-terminal region of Human NgR3/NgRH2/RTN4RL1 (NP_848663). Peptide sequence RPGHRKPGKNCTNPRNRNQISKAGAGKQAPELPDYAPDYQHKFSFDIMPT
100 μg
Primary
Human
Purified
Western Blot
Unconjugated
PBS buffer, 2% sucrose
Rabbit
Affinity purified
RUO
146760
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.