Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NGRN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559537
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559537 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559537 Supplier Invitrogen™ Supplier No. PA559537
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGPIGREPDP DSDWEPEERE LQEVESTLKR QKQAIRFQKI RRQMEAPGAP PRTLTWEAME QIRYLHEEFP ESWSVPRLAE G Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.

NGRN may be involved in neuronal differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NGRN
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NGRN
Gene Accession No. Q9NPE2
Gene Alias AW552001; DSC92; FI58G; FI58Gm; HT020; mesenchymal stem cell protein DSC92; m-Neugrin; Neugrin; neugrin, neurite outgrowth associated; neurite outgrowth associated protein; neurite outgrowth-associated protein; NGRN; Spinal cord-derived protein FI58G
Gene Symbols NGRN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NGRN. Recombinant protein control fragment (Product #RP-98167).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51335
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.