Learn More
Invitrogen™ NGRN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588367
Description
Immunogen sequence: QAIRFQKIRR QMEAPGAPPR TLTWEAMEQI RYLHEEFPES WSVPRLAEGF DVSTDVIRRV LKSKFLPTLE QKLKQDQKVL KKAGLAHSLQ HLRGSGNTSK LLPAGHSVSG SLLMPGHEAS SKDPNHSTAL KVIESDTHRT NTPRRRKGRN KEIQDLEESF VPVAAPLGHP RELQKYSSDS ESPRGTGSGA LPSGQKLEEL KAEEPDNFSS KVVQRGREFF DSNGNFLYRI; Positive Samples: U-87MG, A-549, 293T, HeLa; Cellular Location: Nucleus.
NGRN may be involved in neuronal differentiation.
Specifications
NGRN | |
Polyclonal | |
Unconjugated | |
NGRN | |
AW552001; DSC92; FI58G; FI58Gm; HT020; mesenchymal stem cell protein DSC92; m-Neugrin; Neugrin; neugrin, neurite outgrowth associated; neurite outgrowth associated protein; neurite outgrowth-associated protein; NGRN; Spinal cord-derived protein FI58G | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51335 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9NPE2 | |
NGRN | |
Recombinant fusion protein containing a sequence corresponding to amino acids 62-291 of human NGRN (NP_0010282602). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.