Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NGRN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103958
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103958 Supplier Invitrogen™ Supplier No. PA5103958
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NGRN.

NGRN may be involved in neuronal differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NGRN
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NGRN
Gene Accession No. Q9NPE2
Gene Alias AW552001; DSC92; FI58G; FI58Gm; HT020; mesenchymal stem cell protein DSC92; m-Neugrin; Neugrin; neugrin, neurite outgrowth associated; neurite outgrowth associated protein; neurite outgrowth-associated protein; NGRN; Spinal cord-derived protein FI58G
Gene Symbols NGRN
Host Species Rabbit
Immunogen A synthesized peptide derived from human NGRN(Accession Q9NPE2), corresponding to amino acid residues V17-Q67.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51335
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.