Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NGX6 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298040UV
Description
NGX6 Polyclonal specifically detects NGX6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
NGX6 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
C9orf127, chromosome 9 open reading frame 127, NAG-5, nasopharyngeal carcinoma expressed 6, nasopharyngeal carcinoma related protein, Nasopharyngeal carcinoma-associated gene 6 protein, NGX6MGC120460, Protein NAG-5, Protein NGX6, RP11-112J3.10, transmembrane protein 8B | |
Produced in rabbits immunized with E. coli-derived Human NGX6 fragment. | |
0.1 mL | |
Cell Cycle and Replication | |
51754 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction