Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGX6 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen NGX6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP160059
SDP
View Documents
Novus Biologicals
NBP160059
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

NGX6 Polyclonal specifically detects NGX6 in Human samples. It is validated for Western Blot.
Specifications

Specifications

NGX6
Polyclonal
Purified
RUO
Q5TCW5
51754
Synthetic peptides corresponding to NGX6 The peptide sequence was selected from the C terminal of NGX6. Peptide sequence FLLPPRAKTDHGVPSGARARGCGYQLCINEQEELGLVGPGGATVSSICAS.
Primary
Western Blot
Unconjugated
Rabbit
Cell Cycle and Replication
C9orf127, chromosome 9 open reading frame 127, NAG-5, nasopharyngeal carcinoma expressed 6, nasopharyngeal carcinoma related protein, Nasopharyngeal carcinoma-associated gene 6 protein, NGX6MGC120460, Protein NAG-5, Protein NGX6, RP11-112J3.10, transmembrane protein 8B
TMEM8B
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.