Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NGX6 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 215931AP150UL

Description
The down-regulation of C9orf127 may be closely associated with tumorigenesis and metastasis of colorectal carcinoma. However, it may not contribute to the development and progression of gastric carcinoma.Specifications
NGX6 | |
Polyclonal | |
Unconjugated | |
TMEM8B | |
C9orf127, NAG 5, NGX6, Protein NAG 5, Protein NGX6, RP11 112J3.10, TMEM8B, Transmembrane protein 8B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51754 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A6NDV4 | |
TMEM8B | |
NGX6 Fusion Protein Ag16167 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction