Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[NH2Me2][(RuCl((R)-tolbinap))2(mu-Cl)3], TCI America™
SDP

Catalog No. N10171G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 749935-02-2
Molecular Formula C98H94Cl5NP4Ru2+2
Molecular Weight (g/mol) 1789.122
MDL Number MFCD09753034
InChI Key KXTUTRXHNWQYSZ-UHFFFAOYSA-M
Synonym Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl]diruthenate(II)
PubChem CID 131674756
IUPAC Name trichloranium;[1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane;N-methylmethanamine;ruthenium;chloride;hydrochloride
SMILES CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.CNC.[ClH2+].[ClH2+].[ClH2+].Cl.[Cl-].[Ru].[Ru

Specifications

Quantity 1 g
Formula Weight 1783.07
Physical Form Crystalline Powder
Chemical Name or Material [NH2Me2][(RuCl((R)-tolbinap))2(mu-Cl)3]
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.