Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NHE3/SLC9A3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | NHE3/SLC9A3 |
---|---|
Concentration | 1.0 mg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
NHE3/SLC9A3 Polyclonal antibody specifically detects NHE3/SLC9A3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
NHE3/SLC9A3 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3 | |
Partial synthetic peptide made to an internal portion of the human NHE3 protein (between amino acids 600-660) [UniProt P48764] | |
Immunogen affinity purified |
1.0 mg/mL | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
PBS | |
6550 | |
Primary | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title