Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHE3/SLC9A3, p Ser552 Antibody (14D5), Novus Biologicals™
SDP

Catalog No. p-5000215 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11081529T 0.025 mL
NB11081529 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11081529T Supplier Novus Biologicals Supplier No. NB11081529SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 15 publications

NHE3/SLC9A3 Monoclonal specifically detects NHE3/SLC9A3 in Human, Mouse, Rat, Porcine, Opossum, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockout Validated, Knockdown Validated.

Specifications

Antigen NHE3/SLC9A3 (p Ser552)
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoassay, KnockDown
Classification Monoclonal
Clone 14D5
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-5 ug/ml, Simple Western 1:1000, Flow Cytometry reported in scientific literature (PMID 21255452), ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunoprecipitation 1:10-1:500, Knockout Validated reported in scientific literature (PMID 31276916), KnockDown Validated reported in scientific literature (PMID 31276916)
Gene Alias isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3
Gene Symbols SLC9A3
Host Species Mouse
Immunogen Synthetic peptide made to a region surrounding the Serine 552 portion of rat NHE3. [UniProt# P26433]
Molecular Weight of Antigen 85 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6550
Test Specificity This antibody detects NHE3 posphorylated at Serine 552, and does not cross-react with the non-phosphopeptide.
Target Species Human, Mouse, Rat, Pig, Opossum, Primate, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.