Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NHE3/SLC9A3, p Ser552 Antibody (14D5), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11081529UV
Description
NHE3/SLC9A3 Monoclonal specifically detects NHE3/SLC9A3 in Human, Mouse, Rat, Porcine, Opossum, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated, Knockdown Validated.Specifications
NHE3/SLC9A3 (p Ser552) | |
Monoclonal | |
DyLight 350 | |
isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3 | |
Mouse | |
85 kDa | |
0.1 mL | |
Lipid and Metabolism, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
6550 | |
Human, Mouse, Rat, Pig, Opossum, Primate, Rabbit | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay, KnockDown | |
14D5 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated, Knockdown Validated | |
SLC9A3 | |
Synthetic peptide made to a region surrounding the Serine 552 portion of rat NHE3. [UniProt# P26433] | |
Protein G purified | |
RUO | |
Primary | |
This antibody detects NHE3 posphorylated at Serine 552, and does not cross-react with the non-phosphopeptide. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction