Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NHE3/SLC9A3, p Ser552 Antibody (14D5), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11081529MFV500
Description
NHE3/SLC9A3 Monoclonal antibody specifically detects NHE3/SLC9A3 in Human, Mouse, Rat, Porcine, Opossum, Primate, Rabbit samples. It is validated for Flow CytometrySpecifications
NHE3/SLC9A3 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Opossum, Primate, Rabbit | |
Purified |
Flow Cytometry | |
14D5 | |
Flow Cytometry | |
isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3 | |
Synthetic peptide made to a region surrounding the Serine 552 portion of rat NHE3. [UniProt# P26433] | |
0.1 mL | |
Lipid and Metabolism, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
6550 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction