Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NHE3/SLC9A3, p Ser605 Antibody (10A8), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11074678AF594
Description
NHE3/SLC9A3 Monoclonal specifically detects NHE3/SLC9A3 in Mouse, Rat, Porcine, Opossum, Primate, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
NHE3/SLC9A3 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
SLC9A3 | |
Synthetic peptide made to a region surrounding the Serine 605 portion of rat NHE3. [UniProtNo. P26433] | |
Primary | |
This antibody detects NHE3 posphorylated at serine 605, and does not cross-react with the non-phosphopeptide. | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
10A8 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3 | |
Mouse | |
0.1 mL | |
6550 | |
Mouse, Rat, Pig, Opossum, Primate, Rabbit | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction