Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NHE3/SLC9A3, p Ser605 Antibody (10A8), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11074678AF700
Description
NHE3/SLC9A3 Monoclonal specifically detects NHE3/SLC9A3 in Mouse, Rat, Porcine, Opossum, Primate, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
NHE3/SLC9A3 (p Ser605) | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SLC9A3 | |
Synthetic peptide made to a region surrounding the Serine 605 portion of rat NHE3. [UniProt# P26433] | |
Protein G purified | |
RUO | |
Primary | |
This antibody detects NHE3 posphorylated at serine 605, and does not cross-react with the non-phosphopeptide. | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, Immunocytochemistry, Immunofluorescence | |
10A8 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3 | |
Mouse | |
85 kDa | |
0.1 mL | |
Lipid and Metabolism, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
6550 | |
Mouse, Rat, Pig, Opossum, Primate, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction