Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NHE3/SLC9A3, p Ser605 Antibody (10A8), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications
$206.50 - $499.50
Specifications
| Antigen | NHE3/SLC9A3 (p Ser605) |
|---|---|
| Clone | 10A8 |
| Concentration | 2.1 mg/mL |
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:20, Immunoprecipitation 1:10-1:500 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11074678T
![]() |
Novus Biologicals
NB11074678SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB11074678
![]() |
Novus Biologicals
NB11074678 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
NHE3/SLC9A3 Monoclonal specifically detects NHE3/SLC9A3 in Mouse, Rat, Porcine, Opossum, Primate, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| NHE3/SLC9A3 (p Ser605) | |
| 2.1 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction | |
| isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3 | |
| SLC9A3 | |
| IgG2b | |
| Protein G purified | |
| This antibody detects NHE3 posphorylated at serine 605, and does not cross-react with the non-phosphopeptide. |
| 10A8 | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:20, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat, Pig, Opossum, Primate, Rabbit | |
| 6550 | |
| Synthetic peptide made to a region surrounding the Serine 605 portion of rat NHE3. [UniProt# P26433] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 85 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title