Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NHE8 Antibody (7A11), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 4 publications
$206.50 - $499.50
Specifications
Antigen | NHE8 |
---|---|
Clone | 7A11 |
Concentration | 1.2 mg/mL |
Dilution | Western Blot 2 ug/ml (ECL), Immunocytochemistry/Immunofluorescence |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11062091T
![]() |
Novus Biologicals
NB11062091SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB11062091
![]() |
Novus Biologicals
NB11062091 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
NHE8 Monoclonal specifically detects NHE8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
NHE8 | |
1.2 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cellular Markers, Golgi Apparatus Markers, Lipid and Metabolism | |
Q9Y2E8 | |
23315 | |
A fusion protein containing the C-terminal 89 residues of human NHE8. [Swiss-Prot# Q9Y2E8] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
80 kDa |
7A11 | |
Western Blot 2 ug/ml (ECL), Immunocytochemistry/Immunofluorescence | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
DKFZp686C03237, KIAA0939DKFZp686C03237, MGC138418, Na(+)/H(+) exchanger 8, NHE-8, NHE8FLJ42500, sodium/hydrogen exchanger 8, solute carrier family 9 (sodium/hydrogen exchanger), solute carrier family 9 (sodium/hydrogen exchanger), isoform 8, solute carrier family 9 (sodium/hydrogen exchanger), member 8, Solute carrier family 9 member 8 | |
SLC9A8 | |
IgG | |
Protein G purified | |
This is specific for the 85 kDa mature glycosylated form of the protein. It does not recognize the 55 kDa immature form. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title